Learn More
Mitegen Quick Lid Grid Box Lid By Nanosoft, Stainless Steel, for Cryo-EM Sample Protection from Contamination
SDP
Catalog No. 5000495010
Encompass
The Nanosoft Quick Lid is a simple, yet essential, stainless steel lid designed to protect your valuable vitrified samples in a grid box from contamination. Placed securely on top of the grid box, its weight acts as a reliable shield against ice particulates and other airborne contaminants that can compromise your cryo-EM samples. Compatible with any pin lid type grid box.
Catalog No. 50-004-95010 Supplier Mitegen Supplier No. MCEMNSGBL01
The Nanosoft Quick Lid is a simple, yet essential, stainless steel lid designed to protect your valuable vitrified samples in a grid box from contamination. Placed securely on top of the grid box, its weight acts as a reliable shield against ice particulates and other airborne contaminants that can compromise your cryo-EM samples. Compatible with any pin lid type grid box.