Learn More
New England Biolabs, Inc. Quick Ligation™ Kit – 150 reactions
SDP
Catalog No. 50811727
Encompass
The Quick Ligation Kit enables ligation of cohesive end or blunt end DNA fragments in 5 minutes at room temperature. (25C)
Catalog No. 50-811-727 Supplier New England Biolabs, Inc. Supplier No. M2200L
The Quick Ligation Kit enables ligation of cohesive end or blunt end DNA fragments in 5 minutes at room temperature. (25C)