Learn More
New England Biolabs, Inc. Quick Ligation™ Kit – 30 reactions
SDP
Catalog No. 50811728
Encompass
The Quick Ligation Kit enables ligation of cohesive end or blunt end DNA fragments in 5 minutes at room temperature. (25C)
Catalog No. 50-811-728 Supplier New England Biolabs, Inc. Supplier No. M2200S
The Quick Ligation Kit enables ligation of cohesive end or blunt end DNA fragments in 5 minutes at room temperature. (25C)