Learn More
MSC Value Collection Quick Link, 3/8 in, 3500 Lb, 5 Pack
Catalog No. 502869727
Encompass
This product consists of stainless steel quick links designed for securing connections. Each quick link features a substantial load capacity and is constructed from durable material, making it suitable for various demanding applications.
  • Supplied in a 5-quantity, 1-pack
  • Features a 3/8 in diameter
  • Offers a 3,500 Lb load limit
  • Constructed from stainless steel
Catalog No. 50-286-9727 Supplier MSC Supplier No. 53589388
This product consists of stainless steel quick links designed for securing connections. Each quick link features a substantial load capacity and is constructed from durable material, making it suitable for various demanding applications.
  • Supplied in a 5-quantity, 1-pack
  • Features a 3/8 in diameter
  • Offers a 3,500 Lb load limit
  • Constructed from stainless steel