Learn More
New England Biolabs, Inc. Quick-Load Purple 1 kb DNA Ladder – 1.25 ml
SDP
Catalog No. 501047528
Encompass
Quick-Load Purple 1 kb DNA Ladder is a pre-mixed, ready-to-load molecular weight DNA marker containing one tracking dye. Tracking dye does not leave a shadow under UV light. Quick-Load Purple 1 kb DNA Ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 10 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 0.5 to 10 kb. The 3 kb fragment has increased intensity to serve as reference band.
Catalog No. 50-104-7528 Supplier New England Biolabs, Inc. Supplier No. N0552S
Quick-Load Purple 1 kb DNA Ladder is a pre-mixed, ready-to-load molecular weight DNA marker containing one tracking dye. Tracking dye does not leave a shadow under UV light. Quick-Load Purple 1 kb DNA Ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 10 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 0.5 to 10 kb. The 3 kb fragment has increased intensity to serve as reference band.