Learn More
New England Biolabs, Inc. Quick-Load Purple 1 kb Plus DNA Ladder – 3.75 ml
SDP
Catalog No. 501526414
Encompass
Quick-Load Purple 1 kb Plus DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye. Tracking dye does not leave a shadow under UV light. Quick-Load Purple 1 kb Plus DNA Ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands.
Catalog No. 50-152-6414 Supplier New England Biolabs, Inc. Supplier No. N0550L
Quick-Load Purple 1 kb Plus DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye. Tracking dye does not leave a shadow under UV light. Quick-Load Purple 1 kb Plus DNA Ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands.