Learn More
Research Products International Corp Quick Rack Tip Transfer System, Natural, 1250ul, 768/cs
SDP
Catalog No. 501978352
Encompass
QuickRack provides a convenient and efficient method for reloading 96 pipette tips into empty racks. Fits most standard 8 x 12 hole pattern tip racks. Eliminates recycling programs by allowing use of existing racks. Reduces plastic waste and storage space by up to 80%. Unique sealing rings ensure a universal fit with most major brands of pipettors. Beveled pipette tips are manufactured from a low retention resin for optimum accuracy.
Catalog No. 50-197-8352 Supplier Research Products International Corp Supplier No. 248793
QuickRack provides a convenient and efficient method for reloading 96 pipette tips into empty racks. Fits most standard 8 x 12 hole pattern tip racks. Eliminates recycling programs by allowing use of existing racks. Reduces plastic waste and storage space by up to 80%. Unique sealing rings ensure a universal fit with most major brands of pipettors. Beveled pipette tips are manufactured from a low retention resin for optimum accuracy.