Learn More
Research Products International Corp QuickRack Tip Transfer System, Yellow, 200ul, 960 per Case
SDP
Catalog No. 50212602
Encompass
QuickRack provides a convenient and efficient method for reloading 96 pipette tips into empty racks. Fits most standard 8 x 12 hole pattern tip racks. Eliminates recycling programs by allowing use of existing racks. Reduces plastic waste and storage space by up to 80%. Beveled pipette tips have a 200 ul capacity and manufactured from a low retention resin for optimum accuracy. Unique sealing rings ensure a universal fit with most major brands of pipettors.
Catalog No. 50-212-602 Supplier Research Products International Corp Supplier No. 248790
QuickRack provides a convenient and efficient method for reloading 96 pipette tips into empty racks. Fits most standard 8 x 12 hole pattern tip racks. Eliminates recycling programs by allowing use of existing racks. Reduces plastic waste and storage space by up to 80%. Beveled pipette tips have a 200 ul capacity and manufactured from a low retention resin for optimum accuracy. Unique sealing rings ensure a universal fit with most major brands of pipettors.