Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Quick-Read™ Precision Grid Urinalysis Slide

Catalog No. 22171725 Shop All Globe Scientific Products
Change view
Click to view available options
No. per Pack:
100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
22-171-725 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-171-725 Supplier Globe Scientific Supplier No. 3800
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Globe Scientific Quick-Read™ Precision Grid Urinalysis Slide is designed to provide accuracy, uniformity and safety in the microscopic examination of urinary sediment.

  • Made from optically clear acrylic for optimal viewing
  • Consists of ten chambers
  • Each chamber has a grid with 81 well defined, pre-measured squares
  • Each pre-measured square holds a specific volume of urinary sediment which facilitates the convenient and rapid microscopic examination of the cellular elements in each specimen.

Order Info

Box of 100 Slides

Specifications

Sterility Non-sterile
Well Count 10
Type Urinalysis Microscope Slide
No. per Pack 100
Precleaned Yes
Material Acrylic
Color Clear
Description 81 well defined, pre-measured squares
For Use With (Application) Used to accurately count cellular elements in Urine sediment, for microscopic examination
Quantity 100
Writing Surface No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.