Learn More
Perkin Elmer US LLC Quick Release Nut
Catalog No. 502113504
Encompass_Preferred
The Universal Demountable Cell comes standard in various IR Sampling kits. This cell is an assembled cell, allowing you to customize the window material and path length to meet your current needs.
Catalog No. 50-211-3504 Supplier Perkin Elmer US LLC Supplier No. 04821131
Add to Cart
Add to Cart
The Universal Demountable Cell comes standard in various IR Sampling kits. This cell is an assembled cell, allowing you to customize the window material and path length to meet your current needs.