Learn More
Med Vet International Quick Release Tap Handle, J0413, For Sizes 2.7mm-4.5mm, Each
Catalog No. 5000581600
Encompass_Preferred
Quick Release Tap Handle Synthes Fit.Fits all taps 2.7mm, 3.5mm, 4.0mm, and 4.5mm.
Catalog No. 50-005-81600 Supplier Med Vet International Supplier No. JOR413
Quick Release Tap Handle Synthes Fit.Fits all taps 2.7mm, 3.5mm, 4.0mm, and 4.5mm.