Learn More
Zymo Research Corporation Quick-RNA FFPE MiniPrep (50 Preps)
SDP
Catalog No. 501251663
Encompass_Preferred
The unique chemistries of the product have been optimized for maximum recovery of total RNA including small/micro RNA species. Simply deparaffinize tissues using Deparaffinization Solution, digest using Proteinase K, heat to reverse chemical crosslinks, and then purify using Zymo-Spin™ Column technology. The result is high-quality RNA (> 17 nt) that is DNA-free and is ready for RT/qPCR, hybridization, sequencing, etc.
Catalog No. 50-125-1663 Supplier Zymo Research Corporation Supplier No. R1008
The unique chemistries of the product have been optimized for maximum recovery of total RNA including small/micro RNA species. Simply deparaffinize tissues using Deparaffinization Solution, digest using Proteinase K, heat to reverse chemical crosslinks, and then purify using Zymo-Spin™ Column technology. The result is high-quality RNA (> 17 nt) that is DNA-free and is ready for RT/qPCR, hybridization, sequencing, etc.