Learn More
Zymo Research Corporation Quick-RNA™ MicroPrep Kit (50 Preps) w/Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. 50444592
Encompass_Preferred
The Quick-RNA Microprep Kit is designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell (up to 106) and tissue samples (up to 5 mg). The procedure combines a unique buffer system with Zymo-Spin™ column technology to yield high quality total RNA (including small RNAs 17-200 nt) in about 10 minutes. The procedure is simple. Add the provided RNA Lysis Buffer to a sample, and then purify the RNA using the Zymo-Spin™Columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing etc. In addition, the kit can be used for the enrichment of small and large RNAs into separate fractions.
Catalog No. 50-444-592 Supplier Zymo Research Corporation Supplier No. R1050
The Quick-RNA Microprep Kit is designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell (up to 106) and tissue samples (up to 5 mg). The procedure combines a unique buffer system with Zymo-Spin™ column technology to yield high quality total RNA (including small RNAs 17-200 nt) in about 10 minutes. The procedure is simple. Add the provided RNA Lysis Buffer to a sample, and then purify the RNA using the Zymo-Spin™Columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing etc. In addition, the kit can be used for the enrichment of small and large RNAs into separate fractions.