Learn More
Research Products International Corp Quick-RNA MiniPrep Kit, w/Zymo-Spin IIICG Columns, Capped, & Spin-Away Filters, 200 Preps
SDP
Catalog No. 501977027
Encompass
The Quick-RNA Miniprep Kit is one of the most innovative RNA isolation kits available, designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell (up to 107) and tissue samples (up to 50 mg). The procedure combines a unique buffer system with Zymo-Spin column technology to yield high quality total RNA (including small RNAs 17-200 nt) in about 10 minutes. The procedure is simple. Add the provided RNA Lysis Buffer to a sample, and then purify the RNA using the Zymo-Spin Columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing etc. In addition, the kit can be used for the enrichment of small and large RNAs into separate fractions.
Broad Range: Extract total RNA (including small/micro RNAs) from any sample.
DNA-Free: Genomic DNA removal column and DNase I included.
Catalog No. 50-197-7027 Supplier Research Products International Corp Supplier No. ZR1055
The Quick-RNA Miniprep Kit is one of the most innovative RNA isolation kits available, designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell (up to 107) and tissue samples (up to 50 mg). The procedure combines a unique buffer system with Zymo-Spin column technology to yield high quality total RNA (including small RNAs 17-200 nt) in about 10 minutes. The procedure is simple. Add the provided RNA Lysis Buffer to a sample, and then purify the RNA using the Zymo-Spin Columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing etc. In addition, the kit can be used for the enrichment of small and large RNAs into separate fractions.
Broad Range: Extract total RNA (including small/micro RNAs) from any sample.
DNA-Free: Genomic DNA removal column and DNase I included.