Learn More
Zymo Research Corporation Quick-RNA™ MiniPrep Kit (50 Preps) w/Zymo-Spin™ IIICG Columns (Capped) & Spin-Away™ Filters
SDP
Catalog No. 50444596
Encompass_Preferred
The procedure of Quick-RNA™ Miniprep Kit uses unique spin-column technology that results in highquality total RNA (including small/microRNAs) and is ready for Next-Gen Sequencing, RT/qPCR, hybridization, etcThe Quick-RNA™ Kits yields high quality total RNA.High levels of genomic DNA contamination arepresent in the preps from Suppliers Q & P but not withthe Quick-RNA™ Kits. Total RNA was isolated fromhuman epithelial cells.RNA isolated with the Quick-RNA™ Kits is DNA-free.Samples isolated with Supplier Q's kit are provided forcomparison. Total RNA was isolated from 106 humanepithelial cells (with in-column DNase treatments forboth kits). Each amplification curve represents anaverage of three independent isolation experiments.
Catalog No. 50-444-596 Supplier Zymo Research Corporation Supplier No. R1054
The procedure of Quick-RNA™ Miniprep Kit uses unique spin-column technology that results in highquality total RNA (including small/microRNAs) and is ready for Next-Gen Sequencing, RT/qPCR, hybridization, etcThe Quick-RNA™ Kits yields high quality total RNA.High levels of genomic DNA contamination arepresent in the preps from Suppliers Q & P but not withthe Quick-RNA™ Kits. Total RNA was isolated fromhuman epithelial cells.RNA isolated with the Quick-RNA™ Kits is DNA-free.Samples isolated with Supplier Q's kit are provided forcomparison. Total RNA was isolated from 106 humanepithelial cells (with in-column DNase treatments forboth kits). Each amplification curve represents anaverage of three independent isolation experiments.