Learn More
Grainger Cofair Roof Repair Tape, Thermoplastic Polyolefin, Black, 6 in. x 25 ft., 1 ea.
Catalog No. 502785723
Encompass_Preferred
This black thermoplastic polyolefin roof repair tape utilizes a proprietary Steel-Loc adhesive, a blend of synthetic co-polymers and butyl rubber, to instantly stop leaks and provide permanent repairs for a variety of materials. Designed for versatility and durability, it adheres to brick, concrete, and numerous roofing surfaces without the need for primer. It maintains performance from -70°F to 200°F and is guaranteed not to crack, harden, or shrink.
  • Can be installed at temperatures as low as 20°F
  • Repairs skylights, gutters, flashings, RVs, mobile homes, trucks, trailers, garden sheds, pond liners, and tarps
  • Features a thickness of 30 mil
  • Provides a 15-year limited warranty
  • VOC-free formulation
Catalog No. 50-278-5723 Supplier Grainger Supplier No. 198JG1
This black thermoplastic polyolefin roof repair tape utilizes a proprietary Steel-Loc adhesive, a blend of synthetic co-polymers and butyl rubber, to instantly stop leaks and provide permanent repairs for a variety of materials. Designed for versatility and durability, it adheres to brick, concrete, and numerous roofing surfaces without the need for primer. It maintains performance from -70°F to 200°F and is guaranteed not to crack, harden, or shrink.
  • Can be installed at temperatures as low as 20°F
  • Repairs skylights, gutters, flashings, RVs, mobile homes, trucks, trailers, garden sheds, pond liners, and tarps
  • Features a thickness of 30 mil
  • Provides a 15-year limited warranty
  • VOC-free formulation