This roofing tape utilizes a proprietary blend of synthetic co-polymers and butyl rubber to provide an immediate and permanent repair solution for a variety of surfaces. Designed for versatility and durability, the tape remains effective in extreme temperatures and is suitable for both commercial and residential maintenance applications, including roof repairs and sealing gaps or leaks.
- Features proprietary Steel-Loc adhesive for a strong, long-lasting bond
- Installs without the need for primer at temperatures as low as 20°F
- Maintains structural integrity across a service temperature range of -70°F to 200°F
- VOC-free formulation guaranteed not to crack, harden, or shrink over time
- Compatible with diverse materials such as EPDM, TPO, metal, vinyl, and wood
- UV protected and moisture resistant with a permeability of <0.05 perms