Learn More
Grainger Cofair Roofing Tape, Thermoplastic Polyolefin, 4 in. x 10 ft., White, 12/Pk.
Catalog No. 502785492
Encompass_Preferred
This roofing tape is designed for instant, permanent leak repair on a variety of surfaces. Utilizing proprietary Steel-Loc adhesive technology, it is formulated to resist cracking, hardening, and shrinking. The tape is effective for sealing skylights, gutters, flashings, and various vehicle and exterior applications.
  • Features thermoplastic polyolefin construction
  • Installs without primer at temperatures as low as 20°F
  • Maintains adhesion in temperature ranges from -70°F to 200°F
  • Compatible with diverse materials including metal, EPDM, vinyl, wood, concrete, and brick
  • Measures 4 in. wide by 10 ft. long with a 30 mil thickness
  • UV protected for outdoor durability
Catalog No. 50-278-5492 Supplier Grainger Supplier No. 198JG2
This roofing tape is designed for instant, permanent leak repair on a variety of surfaces. Utilizing proprietary Steel-Loc adhesive technology, it is formulated to resist cracking, hardening, and shrinking. The tape is effective for sealing skylights, gutters, flashings, and various vehicle and exterior applications.
  • Features thermoplastic polyolefin construction
  • Installs without primer at temperatures as low as 20°F
  • Maintains adhesion in temperature ranges from -70°F to 200°F
  • Compatible with diverse materials including metal, EPDM, vinyl, wood, concrete, and brick
  • Measures 4 in. wide by 10 ft. long with a 30 mil thickness
  • UV protected for outdoor durability