Learn More
Grainger Cofair Quick Roof™ Ultra Bond Roofing Tape, White, Thermoplastic Polyolefin, 6 in. x 25 ft., 1 ea.
Catalog No. 502785489
Encompass_Preferred
This roofing tape features Steel-Loc Adhesive, a proprietary blend of synthetic co-polymers and butyl rubber designed to stop leaks and permanently repair various roofing materials. Engineered from white thermoplastic polyolefin, it provides a durable, UV-protected seal that can be applied in temperatures as low as 20°F.
  • Maintains integrity across a temperature range of -70°F to 200°F
  • Guaranteed not to crack, harden, or shrink
  • Compatible with diverse surfaces including metal, wood, fiberglass, EPDM, and vinyl
  • Suitable for repairing flashings, skylights, gutters, and pond liners
  • Thickness of 30 mil for robust application
Catalog No. 50-278-5489 Supplier Grainger Supplier No. 198JG4
This roofing tape features Steel-Loc Adhesive, a proprietary blend of synthetic co-polymers and butyl rubber designed to stop leaks and permanently repair various roofing materials. Engineered from white thermoplastic polyolefin, it provides a durable, UV-protected seal that can be applied in temperatures as low as 20°F.
  • Maintains integrity across a temperature range of -70°F to 200°F
  • Guaranteed not to crack, harden, or shrink
  • Compatible with diverse surfaces including metal, wood, fiberglass, EPDM, and vinyl
  • Suitable for repairing flashings, skylights, gutters, and pond liners
  • Thickness of 30 mil for robust application