Learn More
BINDER™ Quick Sample Access
SDP
Catalog No. 5000217513 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Units CB 170
BINDER Units CB 170, CBF 170
BINDER Units CB 260, CBF 260
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-002-17513 BINDER Units CB 170, CBF 170
50-002-17510 BINDER Units CB 170
50-002-17530 BINDER Units CB 260, CBF 260
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-002-17513 Supplier BINDER™ Supplier No. 80122726
Only null left
Add to Cart
Add to Cart
Quick sample access allow to open small section of the incubator with viewing window (13 x 25 cm) in the inner door for quick access
A quick sample access option lets you open only a small section of the incubator instead of the whole chamber, which greatly protects the internal atmosphere and the cells.
This design keeps temperature, CO2, humidity, and (if used) O2 much more stable during routine access, which improves cell health and experimental reproducibility.
  • Opening just one segment limits how much warm, humid, CO2‐rich air escapes, so the incubator recovers to setpoints faster after each door opening.
  • More stable conditions from fewer fluctuations in temperature and gases support more consistent growth of sensitive cells, especially in shared incubators that are opened many times per day.
  • Because each opening affects a smaller volume, the incubator needs less CO2 (and N2 in hypoxic mode) to return to the desired atmosphere, which can cut gas use by 40 to 80% compared with a single large inner door.
  • Lower gas consumption extends cylinder lifetime, reduces cost and workload for changing bottles, and helps meet sustainability goals
Specifications
For Use With (Equipment) BINDER Units CB 170, CBF 170
Type Viewing Window
Dimensions (L x W) 25 x 13 cm
Weight (English) 11.02 lb.
Weight (Metric) 5 kg