Learn More
Zymo Research Corporation Quick SARS-CoV-2 rRT-PCR Kit, 1,000 rxns
SDP
Catalog No. 502037274
Encompass_Preferred
Quick SARS-CoV-2 rRT-PCR Kit is a real-time RT-PCR test intended for the qualitative detection of nucleic acids from SARS-CoV-2 in upper respiratory specimens (such as nasal, nasopharyngeal, mid-turbinate or oropharyngeal swabs), and lower respiratory specimens (such as sputum, tracheal aspirates, and bronchoalveolar lavage) from patients suspected of COVID-19 by their healthcare provider.The Quick SARS-CoV-2 rRT-PCR Kit is intended for use by qualified clinical laboratory personnel specifically instructed and trained in the techniques of real-time RT-PCR assays.The Quick SARS-CoV-2 rRT-PCR Kit is only for use under the Food and Drug Administration’s Emergency Use Authorization.
This item is not returnable. View return policy
Catalog No. 50-203-7274 Supplier Zymo Research Corporation Supplier No. R301110K
Add to Cart
Add to Cart
Request a Quote
Quick SARS-CoV-2 rRT-PCR Kit is a real-time RT-PCR test intended for the qualitative detection of nucleic acids from SARS-CoV-2 in upper respiratory specimens (such as nasal, nasopharyngeal, mid-turbinate or oropharyngeal swabs), and lower respiratory specimens (such as sputum, tracheal aspirates, and bronchoalveolar lavage) from patients suspected of COVID-19 by their healthcare provider.The Quick SARS-CoV-2 rRT-PCR Kit is intended for use by qualified clinical laboratory personnel specifically instructed and trained in the techniques of real-time RT-PCR assays.The Quick SARS-CoV-2 rRT-PCR Kit is only for use under the Food and Drug Administration’s Emergency Use Authorization.