Learn More
Beckman Coulter TUBES QS UC 1 X 3-1/2 (BOX-50)
Catalog No. NC9731106
Encompass_Preferred
Quick-Seal, Ultra-Clear Standard Tube, 39mL, 25x89mm, Quantity: 50/Box
Catalog No. NC9731106 Supplier Beckman Coulter Supplier No. 344326
Quick-Seal, Ultra-Clear Standard Tube, 39mL, 25x89mm, Quantity: 50/Box