Learn More
Bio Rad Laboratories QUICK START BOVINE SERUM ALBUM
Catalog No. 509012361
Encompass
Standard, Bovine Serum Albumin; Bio-Rad; Quick Start; 5 x 2mL vials of 2mg/mL
Catalog No. 50-901-2361 Supplier Bio Rad Laboratories Supplier No. 5000206
Standard, Bovine Serum Albumin; Bio-Rad; Quick Start; 5 x 2mL vials of 2mg/mL