Learn More
Bio Rad Laboratories QUICK START BRADFORD PROTEIN A
Supplier:  Bio Rad Laboratories 5000204
Encompass
Kit 4, Protein Assay; Bio-Rad; Quick Start Bradford; Includes dye reagent 1L, bovine gamma-globulin standard set, 2 sets of 7 standards, 0.125-2.0mg/mL, 2mL
Catalog No. 50-901-0037