Learn More
Nelson Jameson Company QUICK SWABS 250/CS 70200684895
Catalog No. NC0844878
Encompass
3M Quick Swabs reduce labor required to prepare, gather and plate environmental samples, increasing efficiencies. Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant. Sampling swab can be used wet or dry and can target hard to reach areas for increased sampling flexibility. Self-contained sampling swab is easy to use and can reduce need for dilution mixing and pipetting for quick sampling.
Catalog No. NC0844878 Supplier Nelson Jameson Company Supplier No. 4603434 B
3M Quick Swabs reduce labor required to prepare, gather and plate environmental samples, increasing efficiencies. Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant. Sampling swab can be used wet or dry and can target hard to reach areas for increased sampling flexibility. Self-contained sampling swab is easy to use and can reduce need for dilution mixing and pipetting for quick sampling.