Learn More
Cell Biolabs Inc QuickTiter™ Adenovirus Titer Immunoassay Kit, 100 assays
Catalog No. NC9346220
Encompass_Preferred
Traditional plaque forming unit assays (PFU) used to measure adenoviral functional titer are tedious and can take up to 10 days for results. Cell Biolabs' QuickTiter™ Adenovirus Titer Immunoassay Kit measures functional titer in less time (2.5 days vs. 10 days) with greater accuracy than traditional PFU assays. Each kit provides sufficient reagents for 100 assays.
Catalog No. NC9346220 Supplier Cell Biolabs Inc Supplier No. VPK109
Traditional plaque forming unit assays (PFU) used to measure adenoviral functional titer are tedious and can take up to 10 days for results. Cell Biolabs' QuickTiter™ Adenovirus Titer Immunoassay Kit measures functional titer in less time (2.5 days vs. 10 days) with greater accuracy than traditional PFU assays. Each kit provides sufficient reagents for 100 assays.