Learn More
Agilent Technologies InfinityLab Quick Turn capillary, stainless steel fitting
Catalog No. NC1724974
Encompass
InfinityLab Quick Turn capillary, stainless steel, 0.17 x 105 mm, long socket both ends, without fitting.
Catalog No. NC1724974 Supplier Agilent Technologies Supplier No. 55001193
InfinityLab Quick Turn capillary, stainless steel, 0.17 x 105 mm, long socket both ends, without fitting.