Learn More
LICOR Quick Western Kit
Catalog No. NC1670796
Encompass_Preferred
The Quick Western Kit provides a universal detection reagent that can be combined with the primary antibody incubation step, eliminating the need for a secondary antibody incubation step. This reduces the overall time to complete a Western blot and provides the advantages of near-infrared fluorescence detection. The kit also serves as a detection solution for post-immunoprecipitation samples by Western blot because it does not bind to denatured mouse monoclonal or rabbit monoclonal antibodies. The key benefit is the ability to use the same antibody for immunoprecipitation and post-immunoprecipitation detection by Western blot. Kit components include IRDye 680RD Detection Reagent, Intercept (PBS) Blocking Buffer, and a Quick Reference Card
Catalog No. NC1670796 Supplier LICOR Supplier No. 92669100
The Quick Western Kit provides a universal detection reagent that can be combined with the primary antibody incubation step, eliminating the need for a secondary antibody incubation step. This reduces the overall time to complete a Western blot and provides the advantages of near-infrared fluorescence detection. The kit also serves as a detection solution for post-immunoprecipitation samples by Western blot because it does not bind to denatured mouse monoclonal or rabbit monoclonal antibodies. The key benefit is the ability to use the same antibody for immunoprecipitation and post-immunoprecipitation detection by Western blot. Kit components include IRDye 680RD Detection Reagent, Intercept (PBS) Blocking Buffer, and a Quick Reference Card