Learn More
Zymo Research Corporation Quick16S Plus Library Kit V3V4
SDP
Supplier:  Zymo Research Corporation D6421PS4
Encompass_Preferred
TheQuick-16S Plus NGS Library Prep Kit (V3-V4, UDI) is the fastest and simplest NGS library prep targeting the V3-V4 region of the 16S rRNA gene for high-throughput sequencing. The automation-friendly protocol utilizes a single qPCR/PCR for combined targeted amplification and barcode addition using specially designed primers. After pooling by equal volume, a single clean-up of the final library is performed, rather than massive AMPurebead-based clean-ups. Additional library quantification analysis such as TapeStationanalysis or gel electrophoresis are not necessary. With these features, the workflow dramatically reduces the hands-on time of library preparation to only 30 minutes.
Catalog No. 50-286-5908