Learn More
Agilent Technologies QUICKCHANGE LIGHTNING MULTISI
Catalog No. NC9899999
Encompass
QuikChange Lightning Multi Site-Directed Mutagenesis Kit contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.
Catalog No. NC9899999 Supplier Agilent Technologies Supplier No. 210516
Add to Cart
Add to Cart
QuikChange Lightning Multi Site-Directed Mutagenesis Kit contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.