Learn More
Zymo Research Corporation Quick-DNA/RNA Blood Tube Kit (50 Prep)
SDP
Catalog No. 501251702
Encompass_Preferred
The purification procedure uses Zymo-Spin™ column technology. Simply bind the sample onto the Zymo-Spin™ IIICG Column with the aid of reservoirs (vacuum compatible). There is no reagent removal and no pelleting for easy direct whole tube processing.High-quality DNA and/or total RNA from 3 ml whole blood is eluted into 50 l of DNase/RNase-Free Water and is ready for any downstream application including RT/qPCR, sequencing, etc.
Catalog No. 50-125-1702 Supplier Zymo Research Corporation Supplier No. R1151
The purification procedure uses Zymo-Spin™ column technology. Simply bind the sample onto the Zymo-Spin™ IIICG Column with the aid of reservoirs (vacuum compatible). There is no reagent removal and no pelleting for easy direct whole tube processing.High-quality DNA and/or total RNA from 3 ml whole blood is eluted into 50 l of DNase/RNase-Free Water and is ready for any downstream application including RT/qPCR, sequencing, etc.