Learn More
LGC GENOMICS LLC (Lucigen) 1000mL; Simple, rapid extraction of PCR-ready DNA for screening and genotyping type applications from hair follicles, quill-end cells of feathers, tissue-culture cells, buccal cells and other samples
Catalog No. NC2145346
Encompass
QuickExtract DNA Extraction Solution - 1000mLSimple, rapid extraction of PCR-ready DNA for screening and genotyping type applicationsFast: 8 minute extraction protocol for most sample typesSimple: No centrifugation steps of spin columns used to help increase yieldsAutomation-friendly: Simple protocol integrated easily into automated workflowsSafe: Uses only non-toxic reagentsRecommended for rapid, easy sample prep for T7E1 CRISPR mutation detection assays
Catalog No. NC2145346 Supplier LGC GENOMICS LLC Supplier No. QE0901L
Add to Cart
Add to Cart
Request a Quote
QuickExtract DNA Extraction Solution - 1000mLSimple, rapid extraction of PCR-ready DNA for screening and genotyping type applicationsFast: 8 minute extraction protocol for most sample typesSimple: No centrifugation steps of spin columns used to help increase yieldsAutomation-friendly: Simple protocol integrated easily into automated workflowsSafe: Uses only non-toxic reagentsRecommended for rapid, easy sample prep for T7E1 CRISPR mutation detection assays