Learn More
LGC GENOMICS LLC (Lucigen) QuickExtract Plant DNA Extraction SolutionRapid and efficient extraction of PCR-ready genomic DNA from plant leaf samples.
Catalog No. NC2145345
Encompass
QuickExtract Plant DNA Extraction SolutionRapid and efficient extraction of PCR-ready genomic DNA from plant leaf samplesFast: PCR-ready DNA in about 8 minutes for most samplesSimple: No bead-beating, freezing, or grinding of plant leaf materialIncreased Yields: No centrifugation or spin columns used which reduce yieldsCompatible: Extracted DNA is compatible with both real-time and end-point PCRSafe: Non-toxic reagents used throughout the procedure
Catalog No. NC2145345 Supplier LGC GENOMICS LLC Supplier No. QEP70750
QuickExtract Plant DNA Extraction SolutionRapid and efficient extraction of PCR-ready genomic DNA from plant leaf samplesFast: PCR-ready DNA in about 8 minutes for most samplesSimple: No bead-beating, freezing, or grinding of plant leaf materialIncreased Yields: No centrifugation or spin columns used which reduce yieldsCompatible: Extracted DNA is compatible with both real-time and end-point PCRSafe: Non-toxic reagents used throughout the procedure