Learn More
LGC GENOMICS LLC (Lucigen) QuickExtract RNA Extraction Kit, 50 mLRapid extraction of RT-PCR ready RNA from cultured mammalian cells.
Catalog No. NC1438345
Encompass
The QuickExtract™ RNA Extraction Kit is a fast, simple way of preparing RNA for RT-PCR (both end-point and real-time). The single-tube system requires only vortex mixing to lyse the cells, and prepare the RNA for cDNA synthesis.
This item is not returnable. View return policy
Catalog No. NC1438345 Supplier LGC GENOMICS LLC Supplier No. QER090150
The QuickExtract™ RNA Extraction Kit is a fast, simple way of preparing RNA for RT-PCR (both end-point and real-time). The single-tube system requires only vortex mixing to lyse the cells, and prepare the RNA for cDNA synthesis.