Learn More
CYTOGNOS INC Quicklysis, 250 tests
Supplier:  CYTOGNOS INC CYTQL1R
Encompass
An erythrocyte lyse-non-wash solution without fixatives that allows studying leukocyte subpopulations by flow cytometry. Is used in whole blood or bone marrow samples after immunofluorescence staining of the cells, and requires no subsequent washing-centrifugation steps reducing processing time and sample handling.
Catalog No. NC1841145