This rescue kit is designed for efficient, reliable recovery of fallen workers, offering a 400 lb. (181 kg) capacity and a 5:1 lifting ratio for mechanical advantage. The system features an intuitive design that enables rapid setup and deployment during emergency situations, incorporating a backup braking system to ensure safety. Engineered for durability, the hardware meets ASTM fifty-hour salt spray test requirements.
- Includes a backup braking system for enhanced safety
- Features high-grade anodized aluminum pulleys
- Utilizes a 7/16 in. (11 mm) polyamide kernmantle rope with 8,000 lbs. (35.6 kN) minimum tensile strength
- Equipped with an aluminum fiberglass rescue remote connection pole that extends up to 12 ft. (3.6 m)
- Comes with a cross-arm anchorage connector made of 3 in. (76 mm) polyester webbing
- Meets OSHA and ANSI Z359.4 standards