Learn More
Electron Microscopy Sciences Quick-Read™ Microscope Slide
SDP
Catalog No. 501931279
Encompass_Preferred
These slides are especially manufactured to cater to the Microscopic Urinalysis testing. They are made from clear acrylic with 10 chambers. Each chamber has 18 pre-measured circles that hold a determined volume of urinary sediment. The circles are able to be seen as a group at 100x magnification.
Catalog No. 50-193-1279 Supplier Electron Microscopy Sciences Supplier No. 6463302
These slides are especially manufactured to cater to the Microscopic Urinalysis testing. They are made from clear acrylic with 10 chambers. Each chamber has 18 pre-measured circles that hold a determined volume of urinary sediment. The circles are able to be seen as a group at 100x magnification.