Learn More
Alkali Scientific QuickSilver™ Buffer Powder, Tris-Glycine-SDS Buffer Powder, 50 pouches
SDP
Catalog No. 503028268
Encompass_Preferred
QuickSilverª Buffer Powder Packs offer extreme convenience and will change the way you prepare buffers in your lab. They are quick, easy to use, and come in a variety of buffers for Nucleic Acid and Protein Electrophoresis, Western Blotting, ELISA, and Cell Culture. Quick and Economical (and low shipping costs!) Convenient premeasured pouches Each pouch makes 1L of 1X buffer No pH adjustment needed Nuclease and protease free Quality and consistency guaranteed Long shelf life Buffers for Electrophoresis of Nucleic Acids and Proteins: TBE, TAE, Fast Running Buffer, Tris-Glycine-SDS, Tris-MOPS-SDS, Tris-MES-SDS Buffers for Western Blotting, ELISA and Cell Culture: TBS, TBST, PBS, PBST
Catalog No. 50-302-8268 Supplier Alkali Scientific Supplier No. RB1100
QuickSilverª Buffer Powder Packs offer extreme convenience and will change the way you prepare buffers in your lab. They are quick, easy to use, and come in a variety of buffers for Nucleic Acid and Protein Electrophoresis, Western Blotting, ELISA, and Cell Culture. Quick and Economical (and low shipping costs!) Convenient premeasured pouches Each pouch makes 1L of 1X buffer No pH adjustment needed Nuclease and protease free Quality and consistency guaranteed Long shelf life Buffers for Electrophoresis of Nucleic Acids and Proteins: TBE, TAE, Fast Running Buffer, Tris-Glycine-SDS, Tris-MOPS-SDS, Tris-MES-SDS Buffers for Western Blotting, ELISA and Cell Culture: TBS, TBST, PBS, PBST