Learn More
Alkali Scientific QuickSilver PBS-T Buffer Powder, 100 Pouches
SDP
Catalog No. 502389543
Encompass_Preferred
QuickSilver™ Buffer Powder packs offer extreme convenience and will change the way you prepare buffers in your lab. They are quick, and easy to use. Convenient premeasured pouches make 1L of 1X buffer. There is no pH adjusent needed and they offer a long shelf life. They come supplied with preprinterd bottle labels for easy identification. 100 pouches per pack.
Catalog No. 50-238-9543 Supplier Alkali Scientific Supplier No. RB1201
QuickSilver™ Buffer Powder packs offer extreme convenience and will change the way you prepare buffers in your lab. They are quick, and easy to use. Convenient premeasured pouches make 1L of 1X buffer. There is no pH adjusent needed and they offer a long shelf life. They come supplied with preprinterd bottle labels for easy identification. 100 pouches per pack.