Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ QuickSlide™ System Cleaner
SDP

Catalog No. 23001715 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
2 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-001-715 2 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-001-715 Supplier Hardy Diagnostics™ Supplier No. QS4000
Add to Cart
Add to Cart

Clean the GramPRO 1 Crystal Violet, Grams Iodine and Safranin lines from the reagent cannulas throughout the system. Hardy Diagnostics™ QuickSlide™ System Cleaner should be performed once a month or every 600 cycles.

  • Cleans the GramPRO 1 Crystal Violet, Grams Iodine and Safranin lines from the reagent cannulas throughout the system including tubing elbows, pump tubes, stainless fitting ports, machined ports and the photo optics windows on the cuvette
  • The product comes in a box with two 500mL bottles of cleaner
  • Each bottle will accommodate one scrub cycle

Specifications

Description Each bottle will accommodate one scrub cycle
For Use With (Equipment) GramPRO 1, GramPRO 80
Includes Two 500mL bottles of cleaner
Quantity 2 x 500 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.