Learn More
Diagnocine LLC Quick Taq HS DyeMix
SDP
Catalog No. NC1842346
Encompass
Taq-based 2x master mix PCR reagent that contains an electrophoresis dye (BPB; bromophenol blue) and anti-Taq antibodies for hot start PCR. This reagent contains all components for PCR except primers and template DNA. This reagent shows specific and efficient amplification. The amplified products can be directly loaded in the wells of agarose or acrylamide gels.
Catalog No. NC1842346 Supplier Diagnocine LLC Supplier No. TYBDTM10110PK
Taq-based 2x master mix PCR reagent that contains an electrophoresis dye (BPB; bromophenol blue) and anti-Taq antibodies for hot start PCR. This reagent contains all components for PCR except primers and template DNA. This reagent shows specific and efficient amplification. The amplified products can be directly loaded in the wells of agarose or acrylamide gels.