Learn More
Contec Quicktask Loaded Compact Bucket System
Catalog No. 17720589 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Polypropylene Bucket, Stainless Steel Cart
Catalog No. Material
17-720-589 Polypropylene Bucket, Stainless Steel Cart
1 options
Catalog No. 17-720-589 Supplier Contec Supplier No. 2942U3BFQ
Add to Cart
Add to Cart
Component of the Compact Bucket System which is an improved versatile multiple bucket system for use with Contec's flat and curved mops.
  • Autoclavable Compact Bucket System is an all-in-one solution designed to make cleaning and disinfecting easier and more efficient.
  • The cart is designed for use with Contec™ mops such as the VertiKlean™ MAX™, VertiKlean™ MAX™ Sealed Edge, EasyCurve™ and QuickTask™ Mopping Systems.
  • The stainless steel cart is available in multiple configurations to meet the unique needs of your environment.
  • Available with either two or three autoclavable buckets and a utility cart handle.
  • Each cart is fitted with quiet, easy-to-move casters that lock in place when needed.
  • The utility handle allows two small buckets (1.6 gal.) or one large bucket (3.9 gal.) to be positioned on the handle, providing additional storage for extra supplies such as clean mop heads, solution spray bottles, wipes, and other items.
  • The cart frame includes an integrated mop head removal tool on the front for easy, hands-free removal of a VertiKlean MAX or VertiKlean MAX Sealed Edge mop head.
  • An optional hands-free removal tool is also available for both VertiKlean MAX and QuickTask mop heads.
  • Two mop holders with mop brace and handle clasps on either side of the cart keep mops securely positioned during transport and storage.
  • The system is provided with stainless steel debris screens positioned in the base of the clean and rinse buckets to prevent mop contact with settled contaminants.
  • On-cart storage for two mop handles and frames allows convenient storage and keeps mops out of the way.
  • Improved casters and connections significantly reduce noise during use and provide more durable, low-rattle attachments.
  • Compact polypropylene buckets with integral drip caps keep buckets in place during use and prevent drips between the buckets and the floor.
  • Buckets can hold either a curved or flat wringer for curved or flat mops.
  • Integral or optional hands-free mop head removal tools allow easy disposal of used VertiKlean MAX and QuickTask mop heads.
  • The cart is 100% autoclavable and constructed from electropolished stainless steel, safe for autoclave use at 121°C for 30 min.
  • Compatible with most cleanroom disinfectants and cleaners for use in critical environments.
  • The utility handle basket holds two 6 L buckets, providing additional storage for clean mop heads, solution bottles, wipes, and other cleaning supplies.
Specifications
Product Type Bucket and Wringer System
Material Polypropylene Bucket, Stainless Steel Cart