Learn More
Cell Biolabs Inc QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated p24 ELISA), Trial Size, 32 assays
Supplier:  Cell Biolabs Inc VPK107T
Encompass_Preferred
Traditional p24 ELISA Kits greatly overestimate the titer of recombinant lentiviral preps, because they detect a signficant amount of free p24 protein that was overexpressed during lentiviral packaging and does not correspond to a viral particle. As much as 90% of p24 in lentiviral supernatant may be non-viral. Cell Biolabs' QuickTiter™ Lentivirus Titer Kit is a p24 ELISA that uses proprietary technology to eliminate free (non-viral) p24 protein from lentiviral preps that can cause overestimation of titer. This gives you a much more accurate measurement of viral titer, so you know exactly how much virus you are using for transduction. The trial size kit provides sufficient reagents for 32 assays including standards and unknown samples.
Catalog No. 50-673-750