Learn More
Cell Biolabs Inc QuickTiter™ Lentivirus Quantitation Kit, 20 assays
Catalog No. NC9293255
Encompass_Preferred
Cell Biolabs' QuickTiter™ Lentivirus Quantitation Kit provides an ultra-fast 45-60 minute procedure that measures physical titer of purified virus or unpurified supernatant via the viral nucleic acid content. Quantitation is performed on a 96-well fluorescence based plate reader. The kit provides sufficient reagents for 20 assays.
Catalog No. NC9293255 Supplier Cell Biolabs Inc Supplier No. VPK112
Cell Biolabs' QuickTiter™ Lentivirus Quantitation Kit provides an ultra-fast 45-60 minute procedure that measures physical titer of purified virus or unpurified supernatant via the viral nucleic acid content. Quantitation is performed on a 96-well fluorescence based plate reader. The kit provides sufficient reagents for 20 assays.