Learn More
Cell Biolabs Inc QuickTiter™ Lentivirus Quantiation Kit (p24 ELISA), 96 assays
Supplier:  Cell Biolabs Inc VPK108H
Encompass_Preferred
Cell Biolabs' QuickTiter™ HIV Lentivirus Quantitation Kit (HIV-1 p24 ELISA) measures the physical titer of recombinant lentivirus via the p24 core protein of HIV-1. Results are quantified on a standard 96-well microplate reader. The kit provides sufficient reagents for 96 assays including standards and unknown samples.
Catalog No. NC9421029