Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ QuickVue™ Adenoviral conjunctivitis Test Kit
SDP

Catalog No. 23043023 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
10/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-023 10/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-023 Supplier Quidel™ Supplier No. 20324
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The first and only CLIA-waived, point-of-care test that detects all known serotypes of adenoviral conjunctivitis

  • Sterile sampling fleece allows for a small fluid sample to be tested
  • Results in 10 minutes
  • Two-color result
  • All components included in kit
  • Room temperature storage

Specifications

Content And Storage Room temperature (15°C to 35°C / 59°F to 86°F)
Control Sets Positive and negative included
Clia Complexity Waived
Sample Type Tear Fluid
Sensitivity 90%
Quantity 10/Kit
Specificity 0.96
Test Time 10 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.