Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ QuickVue™ At-Home OTC COVID-19 Self-Test
SDP

Catalog No. 23044172 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
2 Tests/Kit
25 Tests/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-172 25 Tests/Kit
23-044-171 2 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-172 Supplier Quidel™ Supplier No. 20398
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Quickvue™ At-Home OTC COVID-19 test can accurately detect the Omicron variant (among others) of the Sars-CoV-2, the potentially deadly virus that can lead to COVID-19.

QuickVue™ At-Home OTC COVID-19 Test lets you get rapid results, in the privacy of your own home. Available over-the-counter, everything you need is in the package and taking the test is simple.

This home test is authorized for nonprescription home use with self-collected (unobserved) direct anterior nasal (NS) swab specimens from individuals aged 14 years and older or with adult-collected anterior NS samples from individuals aged 2 years or older.

The test to be used in individuals within 6-days of symptom onset or individuals without symptoms or other epidemiological reasons to suspect COVID-19 infection when tested twice over three days with at least 24 hours (and no more than 48 hours) between tests.

The test uses a gentle self-collected anterior nasal (nares) swab sample to determine a positive or negative COVID-19 result. The swab is swirled in a tube of reagent solution, then removed, before a test strip is inserted. After ten minutes, you can take the strip out of the tube and see your results.

Specifications

Clia Complexity Waived (FDA, EUA)
Detectable Analytes SARS-CoV-2
Format Kit
Sample Type Nasal Swab
Type At-home COVID Test Kit
Includes Individually wrapped sterile foam swabs, test strips, single-use strips, pre-filled tubes, tube holder, instruction sheet
Quantity 25 Tests/Kit
Test Time 10 mins.

For Use Under an Emergency Use Authorization (EUA) Only. For use with direct anterior nasal swab specimens. For In Vitro Diagnostic Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.