Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel QuickVue™ In-Line™ One-Step Strep A Test
SDP

Catalog No. 23043056 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25 Tests
75 Swabs
Type:
Specimen Collection Swab
Streptococcus A Test
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-043056 75 Swabs Specimen Collection Swab
23-043027 25 Tests Streptococcus A Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-043056 Supplier Quidel Supplier No. 00347
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Reaction units, extraction reagents, throat swabs, package insert, and positive and negative controls

Unique patented in-line extraction method reduces hands-on time and provides quick results

  • CLIA Complexity: Waived
  • CPT Codes: 87880/87880QW
  • One-step lateral flow immunoassay for the detection of Group A streptococcal antigen from throat swab specimens
  • Results within 5 minutes
  • Sensitivity: 92%
  • Specificity: 99%
  • Overall accuracy: 96%
  • Ideal for exam room environments
  • Built-in procedural controls confirm proper test performance
  • Two-color read-out makes results easy to interpret
  • For in vitro diagnostic use

Specifications

Type Specimen Collection Swab
Quantity 75 Swabs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.