Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel QuickVue SARS Antigen Test Kit 25/Kit
SDP

Catalog No. 23044167 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-167 25/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-167 Supplier Quidel Supplier No. 20387
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detects SARS-CoV-2 antigens directly from anterior nares swab specimens. The QuickVue SARS Antigen Test provides accurate and reliable results in 10 minutes.

  • Dipstick test format
  • Results in 10 minutes
  • Two-color result
  • All components included in kit
  • Room temperature storage

Specifications

Certifications/Compliance Emergency Use Authorization (EUA)
Content And Storage 15°C to 30°C
Control Sets Included and available for separate purchase
CE Marker No
Clia Complexity Waived
Detectable Analytes SARS-CoV-2
Sample Type Nasal Swab
Sensitivity 96.6%
Type Rapid Diagnostic
For Use With (Application) Lateral Flow Rapid Antigen
Includes Test and Controls
Packaging Type Kit
Quantity 25/Kit
Specificity 0.993
Test Time 10 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.