Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ QuickVue™ TLI Campylobacter Test Kit
SDP

Catalog No. 23043041 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-041 25/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-041 Supplier Quidel™ Supplier No. 20344

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A rapid membrane enzyme-linked immunosorbent assay for the qualitative detection of a Campylobacter-specific antigen in human fecal specimens

  • Results in 30 minutes compared to culture, which requires 72 hours and is prone to false negative results due to stringent sample or specimen transport and growth requirements
  • Can be performed with fresh, frozen, Cary Blair or C&S samples or specimens
  • Detects C. jejuniand, C. coli, C. upsaliensis, and C. lari
  • For patients with signs and symptoms of gastroenteritis
  • Does not rely on bacterial viability, and can be performed on the bench-top with samples that have been exposed to air
  • The test is intended for use with preserved fecal specimens in transport media and unpreserved fecal specimens

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Moderate
Sample Type Human Fecal
Sensitivity 97.1%
Includes Internal and External Controls
Quantity 25/Kit
Specificity 0.991
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.